Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX7C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103394 Supplier Thermo Scientific Supplier No. PA5103394
Add to Cart
Add to Cart

COX7C Polyclonal Antibody

COX7C Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen COX7C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene COX7C
Gene Accession No. P15954, P17665, P80432
Gene Alias COX7C; Cox7c1; COXVIIc; Cytochrome c oxidase polypeptide VIIc; Cytochrome c oxidase polypeptide VIIIA; cytochrome c oxidase subunit 7C; cytochrome c oxidase subunit 7C, mitochondrial; cytochrome c oxidase subunit VIIc; cytochrome c oxidase, subunit VIIc; cytochrome-c oxidase chain VIIc
Gene Symbols COX7C
Host Species Rabbit
Immunogen A synthesized peptide derived from human COX7C(Accession P15954), corresponding to amino acid residues M1-L43.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100188937, 12867, 1350
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.