Learn More
Invitrogen™ COX8A Polyclonal Antibody

Description
The antibody detects endogenous levels of total COX8A protein.
Specifications
Specifications
| Antigen | COX8A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | COX8A |
| Gene Accession No. | P10176 |
| Gene Alias | COX; COX VIII-L; COX8; COX8-2; COX8A; Cox8l; cytochrome c oxidase polypeptide VIII-liver; Cytochrome c oxidase polypeptide VIII-liver/heart; cytochrome c oxidase subunit 8-2; cytochrome c oxidase subunit 8A; cytochrome c oxidase subunit 8A (ubiquitous); cytochrome c oxidase subunit 8A, mitochondrial; cytochrome c oxidase subunit VIII; cytochrome c oxidase subunit VIIIa; cytochrome c oxidase subunit VIIIA (ubiquitous); VIII; VIII-L |
| Gene Symbols | COX8A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.