Learn More
Invitrogen™ Coxsackievirus A6 VP1 Recombinant Rabbit Monoclonal Antibody (HL3698)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA566170

Description
Coxsackievirus A6 VP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
Coxsackievirus A6 (CVA6) is a human enterovirus associated with herpangina in infants. VP1, VP2, and VP3 all have a compact core structure consisting of a classical, beta-sandwich, jelly-roll fold, which forms the bulk of the capsid and each of these protein subunits also have several loops that decorate the outer surface of the capsid.
Specifications
Coxsackievirus A6 VP1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Western Blot, Immunocytochemistry | |
HL3698 | |
Unconjugated | |
Coxsackievirus | |
Recombinant fragment of Coxsackievirus A6 VP1. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.