Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CP 100356 hydrochloride
GREENER_CHOICESDP

Catalog No. 419350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-935-0 50 mg
41-931-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-935-0 Supplier R&D Systems™ Supplier No. 4193/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. P-gp inhibitor

High affinity P-glycoprotein (P-gp) inhibitor (Ki values are 58 and 94 nM for mouse Pgp1a and Pgp1b isoforms). Inhibits calcein-AM uptake in MDR1-transfected MDCKII cells (IC50 = 0.5 μM) and prazosin transport in BCRP-transfected MDCKII cells (IC50 = 1.5 μM). Displays weak or no inhibitory activity against MRP1, OATP1B1 and several major human P450 enzymes (IC50 > 50 μM).

Chemical Identifiers

CAS 142715-48-8
Molecular Formula C31H37ClN4O6

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 597.1
Purity >98%
Molecular Weight (g/mol) 597.109
Chemical Name or Material CP 100356 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.