Learn More
Tocris Bioscience™ CP 122721 hydrochloride

Description
Specifications
Specifications
| CAS | 145877-52-7 |
| Synonym | (2S,3S)-N-[[2-Methoxy-5-(trifluoromethoxy)phenyl]methyl]-2-phenyl-3-piperidinamine dihydrochloride |
| Quantity | 25 mg |
| Molecular Formula | C20H23F3N2O2.2HCl |
| Formula Weight | 453.33 |
| Purity | 98% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.