Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STERIS™ CP-310 Alkaline Detergent

Catalog No. 17010132
Click to view available options
Size:
5 gal.
55 gal.
300 gal.
Container Type:
Drum
Pail
Tote

CP-310 Alkaline Cleaner is a high-performance detergent for pharmaceutical, biotech, and cosmetic industries. Formulated with low-mercury potassium hydroxide, surfactants, and chelating agents, it effectively removes process residues. Low foaming, suitable for CIP and COP.

CP-310 Alkaline Process and Research Cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

HOW CP-310 WORKS

CP-310 Alkaline Detergent is formulated with a proprietary blend of low-mercury grade potassium hydroxide, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in CIP, COP and manual applications.

WHY CP-310?

  • Productivity - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility

Specifications

Product Type Cleaner
Color-coded No
Form Clear Liquid
pH 12.4
Scent Unscented
Sterility Non-sterile
Container Type Tote
Size 300 gal.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.