Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CP 316819
GREENER_CHOICESDP

Catalog No. 354250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-425-0 50 mg
35-4210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-425-0 Supplier R&D Systems™ Supplier No. 3542/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective glycogen phosphorylase inhibitor

Selective glycogen phosphorylase inhibitor [IC50 values are 0.017 and 0.034 μM against human skeletal muscle glycogen phosphorylase a (huSMGPa) and liver glycogen phosphorylase a (huLGPa) respectively]. Antihyperglycemic agent.

Chemical Identifiers

CAS 186392-43-8
Molecular Formula C21H22ClN3O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 415.87
Purity >98%
Molecular Weight (g/mol) 415.874
Chemical Name or Material CP 316819
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.