Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CP 376395 hydrochloride
GREENER_CHOICE

Catalog No. 321210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-121-0 10 mg
32-125-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-121-0 Supplier Tocris Bioscience™ Supplier No. 3212/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective CRF1 antagonist; binds at an allosteric site

CP 376395 hydrochloride is a potent and selective CRF1 receptor antagonist (Ki values are 12 and >10000 nM for CRF1 and CRF2 receptors respectively). Binds at an allosteric site. Attenuates CRF-induced activation of the HPA axis in vivo following i.v. administration. Orally active.

Specifications

Quantity 10 mg
Product Type CP 376395 hydrochloride
Purity >99%
Molecular Weight (g/mol) 362.94
Inhibitors CP 376395 hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.