Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CP 673451
GREENER_CHOICESDP

Catalog No. 599310 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-931-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 59-931-0 Supplier R&D Systems™ Supplier No. 5993/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective PDGFR inhibitor

CP 673451 is a potent and selective PDGFR inhibitor (IC50 values are 1 and 10 nM for PDGFR-β and -α respectively). Selective for PDGFR kinase over a range of related kinases. Inhibits PDGFR-β phosphorylation in glioblastoma xenografts and PDGF-induced angiogenesis in mice.

Specifications

Chemical Name or Material 1-[2-[5-(2-Methoxyethoxy)-1H-benzimidazol-1-yl]-8-quinolinyl]-4-piperidinamine
CAS 343787-29-1
Quantity 10 mg
Solubility Soluble to 100mM in water and to 100mM in DMSO
Product Line Tocris
Molecular Formula C24H27N5O2
Formula Weight 417.5
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.