Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ CP 690550 citrate
GREENER_CHOICESDP

Catalog No. 455650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-565-0 50 mg
45-561-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-565-0 Supplier R&D Systems™ Supplier No. 4556/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent JAK inhibitor

Potent JAK inhibitor (IC50 values are 1, 20, and 112 nM for JAK3, JAK2 and JAK1 respectively). Orally active immunosuppressant; exhibits efficacy in rodent rheumatoid arthritis models.

Chemical Identifiers

CAS 540737-29-9
Molecular Formula C22H28N6O8

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 504.49
Purity >99%
Molecular Weight (g/mol) 504.5
Chemical Name or Material CP 690550 citrate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.