Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CP 945598 hydrochloride
GREENER_CHOICESDP

Catalog No. 423650 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-365-0 50 mg
42-361-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-365-0 Supplier Tocris Bioscience™ Supplier No. 4236/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

CB1 receptor antagonist

Selective, high affinity CB1 antagonist (Ki values are 0.7 and 0.12 nM in binding and functional assays respectively). Displays low affinity for CB2 receptors (Ki = 7600 nM).

Chemical Identifiers

CAS 686347-12-6
Molecular Formula C25H26Cl3N7O

Specifications

Quantity 50 mg
Formula Weight 546.88
Purity >99%
Molecular Weight (g/mol) 546.881
Chemical Name or Material CP 945598 hydrochloride
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.