Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CP 96345
GREENER_CHOICE

Catalog No. 289310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-931-0 10 mg
28-935-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 28-931-0 Supplier Tocris Bioscience™ Supplier No. 2893/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective NK1 antagonist

Potent and selective non-peptide NK1 receptor antagonist. Attenuates substance P-induced salivary response and inhibits neurogenic inflammation in vivo.

Chemical Identifiers

CAS 132746-60-2
Molecular Formula C28H32N2O

Specifications

Quantity 10 mg
Formula Weight 412.57
Purity >98%
Molecular Weight (g/mol) 412.577
Chemical Name or Material CP 96345
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.