Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CP013 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560581 Supplier Invitrogen™ Supplier No. PA560581
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AERNKDPILH VLRQYLDPAQ RGVRVLEVAS GSGQHAAHFA RAFPLAEWQP SDVDQRCLDS IAATTQAQGL TNVKAPLHLD VTWGWEHWGG ILPQSLDLLL C Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CP013
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene METTL26
Gene Accession No. Q497C3, Q96S19, Q9DCS2
Gene Alias 0610011F06Rik; C16orf13; chromosome unknown open reading frame, human C16orf13; JFP2; LOC100770910; methyltransferase like 26; Methyltransferase-like 26; Mettl26; RGD1307155; RIKEN cDNA 0610011F06 gene; similar to CG18661-PA; UPF0585 protein C16orf13; UPF0585 protein C16orf13 homolog
Gene Symbols METTL26
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CP013. Recombinant protein control fragment (Product #RP-98655).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 302998, 68347, 84326
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.