Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CP045 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559432 Supplier Invitrogen™ Supplier No. PA559432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLSTHLEAEK PLRRYGAVEE TAWKTERLGR NQLDIISMAE TTMMPEEIEL EMAKIQRLRE VLVRRESELR FMMDDIQLCK DIMDLKQEL Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CP045
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C16orf45
Gene Accession No. Q96MC5
Gene Alias bMERB domain-containing protein 1; BMERB1; C16orf45; chromosome 16 open reading frame 45; migration inhibitory protein; MINP; RGD1305733; similar to RIKEN cDNA 2900011O08; uncharacterized protein C16orf45; uncharacterized protein C16orf45 homolog
Gene Symbols C16orf45
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CP045. Recombinant protein control fragment (Product #RP-98716).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89927
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.