Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CP045 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567599
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567599 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567599 Supplier Invitrogen™ Supplier No. PA567599
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CP045
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene C16orf45
Gene Accession No. Q5FVJ5, Q96MC5
Gene Alias bMERB domain-containing protein 1; BMERB1; C16orf45; chromosome 16 open reading frame 45; migration inhibitory protein; MINP; RGD1305733; similar to RIKEN cDNA 2900011O08; uncharacterized protein C16orf45; uncharacterized protein C16orf45 homolog
Gene Symbols C16orf45, RGD1305733
Host Species Rabbit
Immunogen Synthesized peptide derived from human CP045.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360459, 89927
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.