Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CP054 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563876
Description
Immunogen sequence: LAERLFRRAL RPDPSHRAPT LVWRPGGELW IEPMGTARER SEDWYGSAVP LLTDRAPEPP TQVGTLEARA TA Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 81%.

Specifications
CP054 | |
Polyclonal | |
Unconjugated | |
C16orf54 | |
C16orf54; chromosome 16 open reading frame 54; transmembrane protein C16orf54; UNQ9389/PRO34280 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283897 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWD8 | |
C16orf54 | |
Recombinant protein corresponding to Human CP054. Recombinant protein control fragment (Product #RP-99620). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction