Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CP054 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524693
Description
CP054 Polyclonal Antibody for Western Blot, Flow
Specifications
| CP054 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6UWD8 | |
| C16orf54 | |
| KLH conjugated synthetic peptide between 170-198 amino acids from the C-terminal region of human CP054. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| C16orf54 | |
| C16orf54; chromosome 16 open reading frame 54; transmembrane protein C16orf54; UNQ9389/PRO34280 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 283897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction