Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CP070 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559439
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559439 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559439 Supplier Invitrogen™ Supplier No. PA559439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HGATVKRMYI YSGNSLQDTK APMMPLSCFL GNVYAESVDV LRDGTGPAGL RLRLLAAGCG PGLLADAKMR VFERSVYFGD SCQDVLSMLG SPHKVFYKS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

C16orf70 (Chromosome 16 Open Reading Frame 70) is a Protein Coding gene. An important paralog of this gene is TBC1D32. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CP070
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C16orf70
Gene Accession No. Q9BSU1
Gene Alias C16orf6; C16orf70; chromosome 16 open reading frame 70; LIN10; lin-10; lin-10 homolog; UPF0183 protein C16orf70
Gene Symbols C16orf70
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CP070. Recombinant protein control fragment (Product #RP-98565).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80262
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.