Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CP110 Mouse anti-Human, Clone: 1A3C12, Proteintech
SDP

Catalog No. 501736874 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6874 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6874 Supplier Proteintech Group Inc Supplier No. 664481IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GSG2 is required for normal alignment of chromosomes at metaphase.

Specifications

Antigen CP110
Applications Western Blot
Classification Monoclonal
Clone 1A3C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CCP110
Gene Accession No. O43303
Gene Alias Centrosomal protein of 110 kDa, CP110, CP110 protein, KIAA0419
Gene Symbols CCP110
Host Species Mouse
Immunogen CP110 Fusion Protein Ag3489
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9738
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.