Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CP110 Polyclonal Antibody, Bethyl Laboratories

Rabbit Polyclonal Antibody

$151.31

Specifications

Antigen CP110
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-156-0144
Encompass_Preferred
View Documents
Bethyl Laboratories, Inc
A301344AT
20 μL
Each of 1 for $151.31
Only null left
Add to Cart
 
Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

CP110 is necessary for centrosome duplication at different stages of procentiole formation. It acts as a key negative regulator of ciliogenesis in collaboration with CEP97 by capping the mother centriole thereby preventing cilia formation. The diseases asssociated with CP110 inclued Spinocerebellar Ataxia 11 and Townes-Brocks Syndrome.
Specifications

Specifications

CP110
Polyclonal
Liquid
RUO
TBS with 0.1% BSA and 0.09% sodium azide
centriolar coiled coil protein 110kDa, centriolar coiled-coil protein 110kDa, centrosomal protein CP110, centrosomal protein of 110 kDa, Cep110, CP110, KIAA0419
CCP110
IgG
Antigen affinity chromatography
Antibody
Western Blot
Unconjugated
Rabbit
Human
O43303
9738
Between 962 and 1012
Primary
4° C
CCP110
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.