Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CP110 Recombinant Rabbit Monoclonal Antibody (HL2312)
GREENER_CHOICE

Catalog No. FEMA550487
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550487 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550487 Supplier Invitrogen™ Supplier No. MA550487
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

CP110 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

Necessary for centrosome duplication at different stages of procentriole formation. Acts as a key negative regulator of ciliogenesis in collaboration with CEP97 by capping the mother centriole thereby preventing cilia formation (PubMed:17719545 PubMed:17681131, PubMed:23486064). Also involved in promoting ciliogenesis. May play a role in the assembly of the mother centriole subdistal appendages (SDA) thereby effecting the fusion of recycling endosomes to basal bodies during cilia formation. Required for correct spindle formation and has a role in regulating cytokinesis and genome stability via cooperation with CALM1 and CETN2 (PubMed:16760425). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CP110
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2312
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCP110
Gene Accession No. O43303, Q7TSH4
Gene Alias 6330503K22Rik; AA415922; AI427129; AW557948; CCP110; centriolar coiled coil protein 110; centriolar coiled coil protein 110kDa; centriolar coiled-coil protein 110; centriolar coiled-coil protein 110kDa; centriolar coiled-coil protein of 110 kDa; Centriolin; centrosomal protein CP110; centrosomal protein of 110 kDa; CEP1; Cep110; CNTRL; CP110; FAN; Kiaa0419; RGD1310509
Gene Symbols CCP110
Host Species Rabbit
Immunogen Recombinant fragmemt of human CP110.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101565, 361634, 9738
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.