Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TechNiGlove CP400 Series Clean Packed Blue Nitrile Gloves
GREENER_CHOICESDP

Catalog No. 19999474 Shop All TechNiGlove International Inc. Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Superior feel and strength versus a typical nitrile exam glove, using new soft modulus technology

  • The new Clean Pack Rival series is the ideal way to clean your lab area, without the expense of a cleanroom glove
  • Superior feel and strength versus a typical nitrile exam glove, using new soft modulus technology
  • The CP400 replaces the cardboard packaging of a boxed exam glove with our double-bagged clean packaging in order to eliminate messy cardboard shedding

Specifications

Chlorinated Yes
Accelerator Free Yes
Applications Industrial, Laboratory
AQL 1.5
Color Purple
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 4 mil
Finger Thickness (Metric) 0.1 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4 mil
Palm Thickness (Metric) 0.1 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Large
Sterility Non-sterile
Thickness (English) 4 mil
Thickness (Metric) 0.1 mm
FDA 510 (k) Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.