Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPA1 (Carboxypeptidase A1) Monoclonal Antibody (OTI1C1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504523

Description
CPA1, also known as CPA, is a 419 amino acid secreted monomeric protein that is highly expressed in pancreatic tissue. Functioning as a pancreatic exopeptidase, CPA1 uses zinc as a cofactor to catalyze the release of C-terminal amino acids from a variety of proteins, thereby playing a key role in protein digestion and degradation. Via its catalytic activity, CPA1 is also thought to be involved in zymogen (proenzyme) inhibition, probably functioning to block enzyme activation pathways. Abnormal levels of CPA1 are associated with pancreatic cancer, suggesting a possible role in either tumor progression or tumor suppression events.Specifications
CPA1 (Carboxypeptidase A1) | |
Monoclonal | |
0.65 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P15085 | |
CPA1 | |
Full length human recombit protein of human CPA1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1C1 | |
Unconjugated | |
CPA1 | |
CPA | |
Mouse | |
Affinity Chromatography | |
RUO | |
1357 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction