Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPA1 (Carboxypeptidase A1) Monoclonal Antibody (OTI1C1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504523S


Description
CPA1 (Carboxypeptidase A1) Monoclonal antibody specifically detects CPA1 (Carboxypeptidase A1) in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
CPA1 (Carboxypeptidase A1) | |
Monoclonal | |
0.65 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P15085 | |
Mouse | |
Affinity chromatography | |
RUO | |
1357 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI1C1 | |
Unconjugated | |
Cpa1 | |
0910001L12Rik; carboxypeptidase A; carboxypeptidase A1; carboxypeptidase A1 (pancreatic); carboxypeptidase A1, pancreatic; CPA; Cpa1; pancreatic carboxypeptidase A; pancreatic procarboxypeptidase A | |
Full length human recombinant protein of human CPA1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction