Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPA1 Mouse anti-Human, Unconjugated, Clone: CPA1/2711, Abnova™

Catalog No. 89337385 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-385 Supplier Abnova Corporation Supplier No. MAB20888
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CPA1 Monoclonal antibody specifically detects CPA1 in Human samples. It is validated for IHC-P.

Specifications

Antigen CPA1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPA1/2711
Conjugate Unconjugated
Description carboxypeptidase A1 (pancreatic)
Gene CPA1
Gene Alias CPA
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human CPA1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1357
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.