Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPA2 Monoclonal Antibody (OTI3D8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504696

Description
Three different forms of human pancreatic procarboxypeptidase A have been isolated. The encoded protein represents the A2 form, which is a monomeric protein with different biochemical properties from the A1 and A3 forms. The A2 form of pancreatic procarboxypeptidase acts on aromatic C-terminal residues and is a secreted protein.Specifications
CPA2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P19222, P48052, Q504N0 | |
CPA2 | |
Full length human recombit protein of human CPA2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3D8 | |
Unconjugated | |
CPA2 | |
carboxypeptidase A2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1358, 232680, 296959 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction