Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPAMD8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556706
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556706 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556706 Supplier Invitrogen™ Supplier No. PA556706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FPETWIWHCL NISDPSGEGT LSVKVPDSIT SWVGEAVALS TSQGLGIAEP SLLKTFKPFF VDFMLPALII RGEQVKIPLS VYNYMGTCAE VYMKLSVPK Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 42%.

The CPAMD8 gene, known as C3 and pregnancy zone protein-like alpha-2-macroglobulin domain-containing 8, is involved in the development of ocular tissues. It has been linked to anterior segment dysgenesis (ASD) and congenital glaucoma, indicating its critical role in eye development. CPAMD8 functions primarily within the non-pigmented epithelium and might participate in cellular communication and structural integrity during the differentiation of retinal organoids. Loss-of-function mutations in CPAMD8 disrupt normal eye formation, leading to various ocular disorders. Recent advancements, such as the establishment of CPAMD8-GFP reporter human embryonic stem cell lines, enable real-time monitoring of its expression, thereby facilitating exploration of its biological functions in retinal differentiation processes. Bioinformatic evaluations and CRISPR/Cas9 technology have provided insights into CPAMD8's role in eye development, with gene editing approaches helping delineate its involvement in ocular health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPAMD8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPAMD8
Gene Accession No. Q8IZJ3
Gene Alias alpha-2 macroglobulin family protein VIP; C3 and PZP like, alpha-2-macroglobulin domain containing 8; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 8; C3 and PZP-like, alpha-2-macroglobulin domain containing 8; CPAMD8; K-CAP; VIP
Gene Symbols CPAMD8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPAMD8. Recombinant protein control fragment (Product #RP-102590).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.