Learn More
CPC™ AseptiQuik DC Body
Click to view available options
Material:
Polycarbonate
Specifications
Specifications
| Material | Polycarbonate |
| Min. Operating Temperature (Metric) | 4°C |
| Maximum Pressure | 60 psi |
| Max. Operating Temperature (Metric) | 40°C |