Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ CPE 3602 Shoe Covers
Shop All International Enviroguard Systems Products
Click to view available options
Color:
Blue
White
Description
CPE disposable shoe covers help protect shoes and surrounding environments from dust, dirt, grime, and liquids. More durable than polypropylene and help protect against chemicals, dirt, dust, liquid splash, non-hazardous particulates, and even shallow immersion.
- Water-resistant and come with a strong elastic closure
- One size fits most; fits up to a men's size 12 1/2 shoe
- Can be worn in both indoor and outdoor environments
- Possible applications include healthcare, cleanrooms, construction, food processing, contract work, pharmaceuticals, moving services, realtors, and more
Specifications
Specifications
Size | X-Large |
Material | CPE |
Latex Free | Yes |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Closure Type | Elastic Ankle |
Sole Type | Standard |
Packaging Quantity | 1000/Cs. |
Resistance | Dry Particulates, Splash |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction