Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPEB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113055
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113055 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113055 Supplier Invitrogen™ Supplier No. PA5113055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CPEB2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

The protein encoded by this gene is highly similar to cytoplasmic polyadenylation element binding protein (CPEB), an mRNA-binding protein that regulates cytoplasmic polyadenylation of mRNA as a trans factor in oogenesis and spermatogenesis. Studies of the similar gene in mice suggested a possible role of this protein in transcriptionally inactive haploid spermatids. Alternatively spliced transcript variants encoding distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPEB2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CPEB2
Gene Accession No. Q7Z5Q1, Q812E0
Gene Alias A630055H10Rik; Cpeb2; CPEB-2; CPE-binding protein 2; CPE-BP2; cytoplasmic polyadenylation element binding protein 2; cytoplasmic polyadenylation element-binding protein 2; hCPEB-2
Gene Symbols CPEB2
Host Species Rabbit
Immunogen Recombinant Human Cytoplasmic polyadenylation element-binding protein 2 protein (167-308AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132864, 231207, 360949
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.