Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPEB3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981283
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1283 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1283 Supplier Bioss Supplier No. BS11340R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Gene ontology (GO) annotation includes 3′-UTR-mediated mRNA destabilization; cellular response to amino acid stimulus; negative regulation of cytoplasmic translation; negative regulation of transcription by RNA polymerase II; positive regulation of nuclear-transcribed mRNA poly(A) tail shortening.

Specifications

Antigen CPEB3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Cpeb3
Gene Accession No. Q8NE35
Gene Alias 4831444O18Rik; BOS_23321; Cpeb3; CPE-binding protein 3; CPE-BP3; cytoplasmic polyadenylation element binding protein 3; Cytoplasmic polyadenylation element-binding protein 3; hCPEB-3; Kiaa0940; mKIAA0940; RGD1564670
Gene Symbols Cpeb3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CPEB3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22849
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.