Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPEB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558371
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558371 Supplier Invitrogen™ Supplier No. PA558371
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDYGFGVLVQ SNTGNKSAFP VRFHPHLQPP HHHQNATPSP AAFINNNTAA NGSSA Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Sequence-specific RNA-binding protein that binds to the cytoplasmic polyadenylation element (CPE), an uridine-rich sequence element (consensus sequence 5'-UUUUUAU-3') within the mRNA 3'-UTR (PubMed:24990967). RNA binding results in a clear conformational change analogous to the Venus fly trap mechanism (PubMed:24990967). Regulates activation of unfolded protein response (UPR) in the process of adaptation to ER stress in liver, by maintaining translation of CPE-regulated mRNAs in conditions in which global protein synthesis is inhibited. Required for cell cycle progression, specifically for cytokinesis and chromosomal segregation (PubMed:26398195). Plays a role as an oncogene promoting tumor growth and progression by positively regulating translation of t-plasminogen activator/PLAT (PubMed:22138752). Stimulates proliferation of melanocytes (PubMed:27857118). In contrast to CPEB1 and CPEB3, does not play role in synaptic plasticity, learning and memory. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPEB4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPEB4
Gene Accession No. Q17RY0
Gene Alias CPEB4; Cpeb-4; CPE-binding protein 4; Cpe-bp4; cytoplasmic polyadenylation element binding protein 4; cytoplasmic polyadenylation element-binding protein 4; hCPEB-4; KIAA1673; mCPEB-4
Gene Symbols CPEB4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPEB4. Recombinant protein control fragment (Product #RP-96852).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80315
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.