Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPED1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552903
Description
Immunogen sequence: GSRKLTAAAP GAVPHTSTET QASRCKKGFS QDKQCFLLSG NAQETRKVKE SMETHFGSHG RRAILYRPPF YSKTELQLHQ HILTQHGYTV VIAEERLNAG LGPGLLEQGD LGSWDLLICL SSKKAEGTPC ISKEVMCQLG L Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 57%.

Specifications
CPED1 | |
Polyclonal | |
Unconjugated | |
CPED1 | |
C7orf58; cadherin like and PC-esterase domain containing 1; cadherin-like and PC-esterase domain containing 1; cadherin-like and PC-esterase domain-containing protein 1; CPED1; UNQ9432/PRO34713 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79974 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A4D0V7 | |
CPED1 | |
Recombinant protein corresponding to Human CPED1. Recombinant protein control fragment (Product #RP-90063). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction