Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPI-17 alpha Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104217
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104217 Supplier Invitrogen™ Supplier No. PA5104217
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CPI17alpha.

CPI-17 is a phosphorylation-dependent inhibitory protein for smooth muscle myosin phosphate. CPI-17 was originally identified as a PKC-potentiated inhibitory protein of protein phosphatase-1, which is dominantly expressed in smooth muscle. Phosphorylation at Threonine 38, in vitro, by PKC or Rho-kinase enhances the inhibitory potency toward myosin phosphatase. CPI-17 is also phosphorylated at Threonine 38 by protein kinase N and might be involved in the calcium sensitization of smooth muscle contraction as a downstream effector of Rho and/or arachidonic acid. CPI-17 is dually phosphorylated at Serine 12 and Threonine 38 by a MYPT-associated kinase, M110 kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPI-17 alpha
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP1R14A
Gene Accession No. Q91VC7, Q96A00, Q99MC0
Gene Alias 1110001M11Rik; 17 kDa PKC-potentiated inhibitory protein of PP1; 17-kDa PKC-potentiated inhibitory protein of PP1; 17-KDa protein; Cpi; CPI17; CPI-17; PKC-potentiated inhibitory protein of PP1; PPP1INL; PPP1R14A; Protein kinase C-potentiated inhibitor protein of 17 kDa; protein phosphatase 1 regulatory inhibitor subunit 14A; protein phosphatase 1 regulatory subunit 14A; protein phosphatase 1, regulatory (inhibitor) subunit 14A
Gene Symbols PPP1R14A
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPP1R14A(Accession Q96A00), corresponding to amino acid residues S12-E62.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114004, 68458, 94274
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.