Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPI17 alpha Antibody (4H10), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104336AF750
Description
CPI17 alpha Monoclonal specifically detects CPI17 alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CPI17 alpha | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
PPP1R14A | |
Recombinant human PPP1R14A (1-147aa) purified from E. coli | |
Primary | |
Human | |
IgG2a κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
4H10 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
17 kDa PKC-potentiated inhibitory protein of PP1, CPI-1717-kDa PKC-potentiated inhibitory protein of PP1, CPI1717-KDa protein, PKC-potentiated inhibitory protein of PP1, PPP1INL, Protein kinase C-potentiated inhibitor protein of 17 kDa, protein phosphatase 1 regulatory subunit 14A, protein phosphatase 1, regulatory (inhibitor) subunit 14A | |
Mouse | |
0.1 mL | |
94274 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction