Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPLX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106398
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106398 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106398 Supplier Invitrogen™ Supplier No. PA5106398
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CPLX2.

Proteins encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPLX2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CPLX2
Gene Accession No. P84086, P84087, Q6PUV4
Gene Alias 921-L; AI413745; AW492120; complexin 2; complexin II; Complexin-2; CPLX2; CPX II; CPX2; CPX-2; CPX-II; DKFZp547D155; Hfb1; LOW QUALITY PROTEIN: complexin-2; MGC138492; synaphin; synaphin 1; synaphin-1; wu:fc27b12
Gene Symbols CPLX2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CPLX2(Accession Q6PUV4), corresponding to amino acid residues P28-K78.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10814, 116657, 12890
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.