Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPLX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117105
Description
Positive Samples: SH-SY5Y, Mouse eye Immunogen sequence: MDFVMKQALG GATKDMGKML GGEEEKDPDA QKKEEERQEA LRQQEEERKA KHARMEAERE KVRQQIRDKY GLKKKEEKEA EEKAALEQPC EGSLTRPKKA IPAGCGDEEE EEEESILDTV LKYLPGPLQD MFKK.
Proteins encoded by the complexin/synaphin gene family are cytosolic proteins that function in synaptic vesicle exocytosis. These proteins bind syntaxin, part of the SNAP receptor. The protein product of this gene binds to the SNAP receptor complex and disrupts it, allowing transmitter release. Two transcript variants encoding the same protein have been found for this gene.
Specifications
CPLX2 | |
Polyclonal | |
Unconjugated | |
CPLX2 | |
921-L; AI413745; AW492120; complexin 2; complexin II; Complexin-2; CPLX2; CPX II; CPX2; CPX-2; CPX-II; DKFZp547D155; Hfb1; LOW QUALITY PROTEIN: complexin-2; MGC138492; synaphin; synaphin 1; synaphin-1; wu:fc27b12 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10814, 116657, 12890 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P84086, P84087, Q6PUV4 | |
CPLX2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-134 of human CPLX2 (NP_0066411). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction