Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPLX2 Recombinant Rabbit Monoclonal Antibody (021)
GREENER_CHOICE

Catalog No. PIMA529186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529186 Supplier Thermo Scientific Supplier No. MA529186
Add to Cart
Add to Cart

CPLX2 Recombinant Rabbit Monoclonal Antibody (21)

CPLX2 Recombinant Monoclonal Antibody for ICC/IF, Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen CPLX2
Applications Flow Cytometry, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 021
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CPLX2
Gene Accession No. Q6PUV4
Gene Alias 921-L; AI413745; AW492120; complexin 2; complexin II; Complexin-2; CPLX2; CPX II; CPX2; CPX-2; CPX-II; DKFZp547D155; Hfb1; LOW QUALITY PROTEIN: complexin-2; MGC138492; synaphin; synaphin 1; synaphin-1; wu:fc27b12
Gene Symbols CPLX2
Host Species Rabbit
Immunogen Recombinant Human Complexin-2/CPLX2 Protein (Asp2-Lys134).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10814
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.