Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPLX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA524148
Description
This antibody is predicted to react with bovine based on sequence homology.
Positively regulates a late step in synaptic vesicle exocytosis.
Specifications
CPLX3 | |
Polyclonal | |
Unconjugated | |
Cplx3 | |
BC016632; complexin 3; complexin III; Complexin-3; CPLX3; CPX III; CpxIII; CPX-III; ERGIC-53L; ERGL; Lamn1l; lectin, mannose-binding, 1 like; Lman1l; Nbla11589; RGD1560104 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
594855 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8WVH0 | |
Cplx3 | |
KLH conjugated synthetic peptide between 52-81 amino acids from the central region of human CPLX3. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction