Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPLX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532059 Supplier Invitrogen™ Supplier No. PA532059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain, mouse eye. Predicted reactivity: Mouse (95%), Rat (95%), Rhesus Monkey (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Positively regulates a late step in synaptic vesicle exocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPLX3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Cplx3
Gene Accession No. Q8R1B5, Q8WVH0
Gene Alias BC016632; complexin 3; complexin III; Complexin-3; CPLX3; CPX III; CpxIII; CPX-III; ERGIC-53L; ERGL; Lamn1l; lectin, mannose-binding, 1 like; Lman1l; Nbla11589; RGD1560104
Gene Symbols Cplx3
Host Species Rabbit
Immunogen Full length human Complexin 3 Recombinant protein.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235415, 594855
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.