Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPLX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589364
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589364 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589364 Supplier Invitrogen™ Supplier No. PA589364
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAQFTQRKAE RATLRSHFRD KYRLPKNETD ESQIQMAGGD VELPRELAKM IEEDTEEEEE KASVLGQLAS LPGLNLGSLK DKAQATLGDL KQSAEKCHVM; Positive Samples: Mouse brain, Mouse eye, Rat brain; Cellular Location: Cell junction, Lipid-anchor, Membrane, synapse.

Positively regulates a late step in synaptic vesicle exocytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPLX3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Cplx3
Gene Accession No. Q8R1B5
Gene Alias BC016632; complexin 3; complexin III; Complexin-3; CPLX3; CPX III; CpxIII; CPX-III; ERGIC-53L; ERGL; Lamn1l; lectin, mannose-binding, 1 like; Lman1l; Nbla11589; RGD1560104
Gene Symbols Cplx3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 59-158 of human CPLX3 (NP_0010251761).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235415, 501005
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.