Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPLX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109582 Supplier Invitrogen™ Supplier No. PA5109582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAFLMKSMIS NQVKNLGFGG GSEENKEEGG ASDPAAAQGM TREEYEEYQK QMIEEKMERD AAFTQKKAER ACLRVHLREK YRLPKSEMDE NQIQMAGDDV DLPEDLRKMV DEDQEEEEDK DSILGQIQNL QNMDLDTIKE KAQATFTEIK QTAEQKCSVM.

CPLX4 (Complexin 4) is a Protein Coding gene. Diseases associated with CPLX4 includeLeft Bundle Branch Hemiblock. Among its related pathways areSynaptic vesicle cycle. Gene Ontology (GO) annotations related to this gene includesyntaxin bindingandSNARE binding. An important paralog of this gene isCPLX3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPLX4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Cplx4
Gene Accession No. 0
Gene Alias A930004D23Rik; complexin 4; complexin IV; Complexin-4; CPLX4; CPX IV; CPXIV; CPX-IV; Gm957
Gene Symbols Cplx4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human CPLX4 (NP_857637.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361342
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.