Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPM Monoclonal Antibody (OTI1C10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807318

Description
Specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.Specifications
CPM | |
Monoclonal | |
Unconjugated | |
CPM | |
carboxypeptidase M | |
Mouse | |
Affinity Chromatography | |
RUO | |
1368 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI1C10 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P14384 | |
CPM | |
Human recombit protein fragment corresponding to amino acids 124-423 of human CPM produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction