Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101498
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101498 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101498 Supplier Invitrogen™ Supplier No. PA5101498
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CPN1.

CPN1 is a plasma metallo-protease that cleaves basic amino acids from the C terminal of peptides and proteins. The protein is important in the regulation of peptides like kinins and anaphylatoxins, and has also been known as kininase-1 and anaphylatoxin inactivator. This protein is a tetramer comprised of two identical regulatory subunits and two identical catalytic subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPN1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CPN1
Gene Accession No. P15169, Q9JJN5
Gene Alias 0610011F20Rik; ACBP; Anaphylatoxin inactivator; arginine carboxypeptidase; carboxypeptidase K; carboxypeptidase N catalytic chain; carboxypeptidase N catalytic subunit; Carboxypeptidase N polypeptide 1; carboxypeptidase N polypeptide 1 50 kD; carboxypeptidase N small subunit; carboxypeptidase N subunit 1; carboxypeptidase N, polypeptide 1; carboxypeptidase N, polypeptide 1, 50kD; cb1037; CPN; CPN1; fa99g08; FLJ40792; kininase I; kininase-1; Lysine carboxypeptidase; plasma carboxypeptidase B; SCPN; serum carboxypeptidase N; Unknown (protein for MGC:128810); wu:fa99g08; zgc:77485
Gene Symbols CPN1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CPN1(Accession P15169), corresponding to amino acid residues I419-A458.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1369, 93721
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.