Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPN1 Polyclonal Antibody, Huabio

Catalog No. 502544545
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4545 Supplier HUABIO RESEARCH INC Supplier No. HA500462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CPN1 Polyclonal antibody specifically detects CPN1 in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen CPN1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CPN1
Gene Accession No. P15169, Q9JJN5
Gene Alias 0610011F20Rik; ACBP; Anaphylatoxin inactivator; arginine carboxypeptidase; carboxypeptidase K; carboxypeptidase N catalytic chain; carboxypeptidase N catalytic subunit; Carboxypeptidase N polypeptide 1; carboxypeptidase N polypeptide 1 50 kD; carboxypeptidase N small subunit; carboxypeptidase N subunit 1; carboxypeptidase N, polypeptide 1; carboxypeptidase N, polypeptide 1, 50kD; cb1037; CPN; CPN1; fa99g08; FLJ40792; kininase I; kininase-1; Lysine carboxypeptidase; plasma carboxypeptidase B; SCPN; serum carboxypeptidase N; Unknown (protein for MGC:128810); wu:fa99g08; zgc:77485
Host Species Rabbit
Immunogen Recombinant protein within human CPN1 aa 1-200/458.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1369, 93721
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.