Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106633
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106633 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106633 Supplier Invitrogen™ Supplier No. PA5106633
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CPN2.

CPN2, the 83 kDa subunit binds and stabilizes the catalytic subunit at 37 degrees Celsius and keeps it in circulation. Under some circumstances it may be an allosteric modifier of the catalytic subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPN2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CPN2
Gene Accession No. P22792
Gene Alias 1300018K11Rik; ACBP; arginine carboxypeptidase; carboxypeptidase N 83 kDa chain; Carboxypeptidase N large subunit; Carboxypeptidase N polypeptide 2; Carboxypeptidase N regulatory subunit; carboxypeptidase N subunit 2; carboxypeptidase N, polypeptide 2; carboxypeptidase N, polypeptide 2 homolog; carboxypeptidase N, polypeptide 2, 83kD; CPN reg; Cpn2; CPNreg; CPN-reg; RGD1305170
Gene Symbols CPN2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CPN2(Accession P22792), corresponding to amino acid residues R449-C499.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1370
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.