Learn More
Invitrogen™ CPNE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527412
Description
Recommended positive controls: 3xFlag-human CPNE1-transfected 293T. Predicted reactivity: Mouse (93%), Rat (93%), Dog (93%), Pig (95%), Rabbit (92%), Sheep (95%), Rhesus Monkey (97%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a calcium-dependent protein that also contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, the encoded protein does not contain a predicted signal sequence or transmembrane domains. This protein has a broad tissue distribution and it may function in membrane trafficking. This gene and the gene for RNA binding motif protein 12 overlap at map location 20q11.21. Alternate splicing results in multiple transcript variants encoding different proteins.
Specifications
| CPNE1 | |
| Polyclonal | |
| Unconjugated | |
| CPNE1 | |
| 1810028N16Rik; Chromobindin 17; copine 1; copine I; copine III; copine III, like; copine-1; COPN1; CPN1; Cpne1; cpne3; cpne3l; MGC1142; mKIAA4108; OTTHUMP00000030775; OTTHUMP00000030776; Unknown (protein for IMAGE:8126132); wu:fb69d07; wu:fc45a04; zgc:55873 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8904 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.48 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q99829 | |
| CPNE1 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human Copine 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.