Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPNE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541780 Supplier Invitrogen™ Supplier No. PA541780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TVQKLRFGIY DIDNKTPELR DDDFLGGAEC SLGQIVSSQV LTLPLMLKPG Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Sheep: 93%; Zebrafish: 79%.

Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a calcium-dependent protein that also contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, the encoded protein does not contain a predicted signal sequence or transmembrane domains. This protein has a broad tissue distribution and it may function in membrane trafficking. This gene and the gene for RNA binding motif protein 12 overlap at map location 20q11.21. Alternate splicing results in multiple transcript variants encoding different proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CPNE1
Gene Accession No. Q99829
Gene Alias 1810028N16Rik; Chromobindin 17; copine 1; copine I; copine III; copine III, like; copine-1; COPN1; CPN1; Cpne1; cpne3; cpne3l; MGC1142; mKIAA4108; OTTHUMP00000030775; OTTHUMP00000030776; Unknown (protein for IMAGE:8126132); wu:fb69d07; wu:fc45a04; zgc:55873
Gene Symbols CPNE1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CPNE1 (aa 70-119).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8904
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.