Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPNE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115047
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115047 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115047 Supplier Thermo Scientific Supplier No. PA5115047
Add to Cart
Add to Cart

CPNE1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CPNE1
Gene Accession No. Q99829
Gene Alias 1810028N16Rik; Chromobindin 17; copine 1; copine I; copine III; copine III, like; copine-1; COPN1; CPN1; Cpne1; cpne3; cpne3l; MGC1142; mKIAA4108; OTTHUMP00000030775; OTTHUMP00000030776; Unknown (protein for IMAGE:8126132); wu:fb69d07; wu:fc45a04; zgc:55873
Gene Symbols CPNE1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CPNE1(Accession Q99829), corresponding to amino acid residues L497-A537.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8904
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.