Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CPNE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556186 Supplier Invitrogen™ Supplier No. PA556186
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAQCVTKVA LNVSCANLLD KDIGSKSDPL CVLFLNTSGQ QWYEVERTER IKNCLNPQFS KTFIIDYYFE VVQKLKFGVY DIDNK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a protein which contains two type II C2 domains in the amino-terminus and an A domain-like sequence in the carboxy-terminus. The A domain mediates interactions between integrins and extracellular ligands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CPNE3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CPNE3
Gene Accession No. O75131
Gene Alias 5430428M23Rik; 5730450C07Rik; C85108; copine 3; copine III; Copine-3; CPN3; Cpne3; KIAA0636; PRO1071
Gene Symbols CPNE3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CPNE3. Recombinant protein control fragment (Product #RP-93743).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8895
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.